missing translation for 'onlineSavingsMsg'
Learn More

Verteporfin, MedChemExpress

Product Code. 16444257
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444257 50 mg 50mg
16418368 10 mg 10mg
16460788 5 mg 5mg
16428136 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16444257 Supplier Medchem Express Supplier No. HYB0146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Verteporfin (CL 318952) is a photosensitizer for photodynamic therapy to eliminate the abnormal blood vessels in the eye associated with conditions such as age-related macular degeneration. Verteporfin is a YAP inhibitor which disrupts YAP-TEAD interactions. Verteporfin induces cell apoptosis. Verteporfinis an autophagy inhibitor that blocks autophagy at an early stage by inhibiting autophagosome formation.

Verteporfin (CL 318952) is a photosensitizer for photodynamic therapy to eliminate the abnormal blood vessels in the eye associated with conditions such as age-related macular degeneration. Verteporfin is a YAP inhibitor which disrupts YAP-TEAD interactions. Verteporfin induces cell apoptosis. Verteporfinis an autophagy inhibitor that blocks autophagy at an early stage by inhibiting autophagosome formation.

Specifications

Chemical Name or Material Verteporfin
CAS 129497-78-5
Color Brown
Purity Grade Notes Research
Molecular Formula C41H42N4O8
Quantity 50 mg
Synonym CL 318952
Solubility Information DMSO : 5 mg/mL (6.96 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@]1(/C2=C/C(N3)=C4C)C(C(/C=C(C(C)=C/5C=C)\NC5=C/C6=N/C(C(CCC(OC)=O)=C6C)=C\C3=C4CCC(O)=O)=N2)=CC=C(C(OC)=O)[C@H]1C(OC)=O.C[C@]7(/C8=C/C(N9)=C%10C)C(C(/C=C(C(C)=C/%11C=C)\NC%11=C/C%12=N/C(C(CCC(O)=O)=C%12C)=C\C9=C%10CCC(OC)=O)=N8)=CC=C(C(OC)=O)[C@H]7C(OC)=O
Molecular Weight (g/mol) 718.79
Formula Weight 718.79
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.