Learn More
Saint Gobain Life Sciences Versilic™ Silicone Caps
Product Code. 11724638
Click to view available options
Diameter (Metric) Bottom:
6 mm
9 mm
10 mm
94 mm
Unit Size:
Each
Pack of 500
Product Code. 11724638
Brand: Saint Gobain Life Sciences 261016
Only null left
Add to basket
Add to basket
Use these stoppers for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Versilic™ Silicone Caps are particularly suited for all types of tissue culture work.
  • Biologically neutral and non-toxic silicone caps for closing tubes and flasks
  • Odorless, tasteless, non-adherent and resistant to aging
  • Nontoxic, biologically neutral
  • FDA/BGA approved
  • For test tubes
  • Easy to install
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance FDA Compliant Food Contact, BFR, BGA
Quantity 500 Pk.
Temperature Range (Metric) -70 to +250°C
Autoclavable Autoclavable
Diameter (Metric) Bottom 9 mm
Diameter (Metric) Top 16 mm
Material Silicone
Product Type Silicone Stopper