Learn More
Ansell VersaTouch™ 92-205 Nitrile Disposable Glove
Shop All Ansell Products
Click to view available options
:
Size: 2X-Large
Description
- A thin and lightweight nitrile glove with textured fingertips for precise handling and enhanced tactility
- Ideal for a wide range of food handling applications
Specifications
Specifications
| Certifications/Compliance (Europe) | Category III|EC 1935/2004|EC 2023/2006|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|FDA21 CFR 177-2600-US Food Contact Approved|ISO 13485|ISO 9001 |
| Chlorinated | No |
| Applications | Industrial, Laboratory |
| Color | White |
| Finger Thickness (English) | 3.9 mil |
| Finger Thickness (Metric) | 0.1 mm |
| Latex Free | Yes |
| Material | Nitrile |
| Palm Thickness (English) | 2.8 mil |
| Palm Thickness (Metric) | 0.07 mm |
| Show More |