Learn More
Thermo Scientific™ VersaRack for VersaCool™ Refrigerated Circulating Bath, Stainless steel, adjustable
Product Code. 15479529 Shop All Thermo Scientific Products
Click to view available options
Type:
Tube Rack
Unit Size:
Each
Product Code. 15479529
Brand: Thermo Scientific™ 230000001
Only null left
Add to basket
Add to basket
Stainless steel adjustable rack. For use with Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Utilize the VersaRack for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
With the ability to accommodate a wide range of beaker and test tube sizes, the VersaRack offers a tool-less adjustable height setting . Its drip-less features allows it to rests over the bath opening saving you valuable bench space.
TRUSTED_SUSTAINABILITY
Specifications
Description VersaRack - Stainless steel adjustable rack
For Use With (Equipment) Thermo Scientific VersaCool Refrigerated Circulating Bath
Type Tube Rack
Holds Holds up to 75 test Tubes that are 10 mm dia.
Holds up to 48 test Tubes that are 16 mm dia.
Holds up to 21 test Tubes that are 25 mm dia.