Learn More
3M™ Versaflo™ S-Series Headcovers and Hoods
Product Code. 13481177
Click to view available options
Product Type:
Hood with Faceseal
Hood with Inner Shroud
Hood with Neck Collar
Painter's Hood with Inner Shroud Assembly
Respirator Hood
Color:
Gray
White
Model:
S-433 L
S-433 L-5
S-433 S
S-605-10
S-607-10
S-655
S-657
S-707-10
S-757
S-805-5
S-855E
Unit Size:
Each
Pack of 10
Pack of 5
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 13481177
Brand: 3M™ 7100079873
Only null left
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
3M™ Versaflo™ S-Series Headcovers and Hoods offer a variety of options for faceseals, collars, shrouds, and suspensions, allowing you to choose the respiratory protection that provides the most value and comfort for your team.
  • S-Series respiratory headcovers and hoods are specifically designed to work together with your Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
  • Provides head, face, neck and shoulder coverage from dust and paint spray.
  • Provides a good field of view.
  • Helps protect eyes and face from liquid splash and low energy flying particles and dust.
  • Versatile accessories available that can help you adapt the hood to your particular environment.
TRUSTED_SUSTAINABILITY
Specifications
Assigned Protection Factor (APF) 1000
Certifications/Compliance NIOSH
Description Premium Replacement Hood with Sealed Seams and Inner Collar, S-805-5
Color Gray
Material Zytron 200
Product Type Hood with Neck Collar
For Use With (Application) Assembly, facility maintenance, machine operation, painting, sanding
For Use With (Equipment) GVP, Breathe-Easy™, V Series, T-300, T-600, T-800
Includes Hood with neck collar only
Model S-805-5
Quantity 5/Cs.
Suspension Type Premium Resusable Suspension
Visor Type Acetate