missing translation for 'onlineSavingsMsg'
Learn More

Vernakalant Hydrochloride, MedChemExpress

Product Code. p-200098028
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Product Code. Quantity unitSize
16492127 50 mg 50mg
16486406 100 mg 100mg
16445988 10 mg 10mg
16438607 5 mg 5mg
16400216 2 mg 2mg
5 options
This item is not returnable. View return policy

Product Code. 16492127

Brand: Medchem Express HY14183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vernakalant hydrochloride is a mixed voltage- and frequency-dependent Na+ and atria-preferred K+ channel blocker. IC50 for block by Vernakalant of wild-type and mutant Kv1.5 channels Fractional block is 13.35±0.93 μM, 0.61±0.03 μM, and 1.63±0.09 μM for Kv1.5 channelwt, Kv1.5 channelI508F, Kv1.5 channelT479A, respectively.

Vernakalant hydrochloride is a mixed voltage- and frequency-dependent Na+ and atria-preferred K+ channel blocker. IC50 for block by Vernakalant of wild-type and mutant Kv1.5 channels Fractional block is 13.35±0.93 μM, 0.61±0.03 μM, and 1.63±0.09 μM for Kv1.5 channelwt, Kv1.5 channelI508F, Kv1.5 channelT479A, respectively.

Specifications

Chemical Name or Material Vernakalant Hydrochloride
CAS 748810-28-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H32ClNO4
Quantity 50 mg
Synonym RSD1235 hydrochloride
Solubility Information DMSO : 50 mg/mL (129.56 mM; Need ultrasonic) ∣H2O : 50 mg/mL (129.56 mM; Need ultrasonic)
SMILES COC1=C(OC)C=C(CCO[C@H]2[C@H](N3CC[C@@H](O)C3)CCCC2)C=C1.Cl
Molecular Weight (g/mol) 385.93
Formula Weight 385.93
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.