missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Veriti™ Thermal Cycler, IQ User-Performed Test Procedures

Product Code. 15131766 Shop All Applied Biosystems Products
Click to view available options
:
1 each
This item is not returnable. View return policy

Product Code. 15131766

Brand: Applied Biosystems™ 4413751

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Veriti™ Thermal Cycler, IQ User-Performed Test Procedures

In laboratories regulated typically by GxP and ISO, in compliance with government and international standards requires documented verification that your systems are installed and functioning according to their operational specifications. This IQ (Installation Qualification) User-Performed Test Procedures package provides you with test procedures to document that your Veriti™ Thermal Cycler is installed to manufacturer’s specifications.

Procedures should be performed when instrument has been relocated or modifications have been made to the instrument’s configuration.

Package includes a bound, printed copy of test procedures. Directions for performing the test procedures can be found in the instrument user manual. Package does not include Temperature Verification Kits and Probes.

For Research Use Only. Not for use in diagnostics procedures.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Procedures and instructions for initial qualification of Veriti™ Thermal Cycler
Product Type Veriti™ Thermal Cycler IQ User-Performed Test Procedures
For Use With (Application) Genotyping and Genomic Profiling, PCR
Description Veriti™ Thermal Cycler IQ User-Performed Test Procedures, 1 Each, Room Temperature Shipping Condition, Use for Genotyping and Genomic Profiling, PCR, Veriti™ Thermal Cycler is Installed to Manufacturer's Specifications
Quantity 1 Ea.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.