missing translation for 'onlineSavingsMsg'
Learn More

Verinurad, MedChemExpress

Product Code. p-200096475
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493187

Brand: Medchem Express HY16733

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Verinurad (RDEA3170) is a highly potent and specific URAT1 inhibitor with an IC50 of 25 nM.

Verinurad (RDEA3170) is a highly potent and specific URAT1 inhibitor with an IC50 of 25 nM.

Specifications

Chemical Name or Material Verinurad
CAS 1352792-74-5
Color White
Purity Grade Notes Research
Molecular Formula C20H16N2O2S
Quantity 50 mg
Synonym RDEA3170
Solubility Information DMSO : ≥ 30 mg/mL (86.10 mM)
SMILES CC(C)(SC1=C(C2=C3C=CC=CC3=C(C#N)C=C2)C=NC=C1)C(O)=O
Molecular Weight (g/mol) 348.42
Formula Weight 348.42
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.