missing translation for 'onlineSavingsMsg'
Learn More

Verapamil hydrochloride, MedChemExpress

Product Code. p-200096808
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
16412009 1 g 1g
16440718 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 16412009

Brand: Medchem Express HYA0064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Verapamil hydrochloride ((±)-Verapamil hydrochloride) is a calcium channel blocker and a potent and orally active first-generation P-glycoprotein (P-gp) inhibitor. Verapamil hydrochloride also inhibits CYP3A4. Verapamil hydrochloride has the potential for high blood pressure, heart arrhythmias and angina research.

Verapamil hydrochloride ((±)-Verapamil hydrochloride) is a calcium channel blocker and a potent and orally active first-generation P-glycoprotein (P-gp) inhibitor. Verapamil hydrochloride also inhibits CYP3A4. Verapamil hydrochloride has the potential for high blood pressure, heart arrhythmias and angina research.

Specifications

Chemical Name or Material Verapamil hydrochloride
CAS 152-11-4
Color White
Purity Grade Notes Research
Molecular Formula C27H39ClN2O4
Quantity 1 g
Synonym (±)-Verapamil hydrochlorideCP-16533-1 hydrochloride
Solubility Information H2O : 50 mg/mL (101.82 mM; Need ultrasonic) ∣DMSO : ≥ 31 mg/mL (63.13 mM)
SMILES [H]Cl.COC1=CC=C(C(C#N)(C(C)C)CCCN(CCC2=CC=C(OC)C(OC)=C2)C)C=C1OC
Molecular Weight (g/mol) 491.06
Formula Weight 491.06
Percent Purity 99.01%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.