missing translation for 'onlineSavingsMsg'
Learn More

Velneprit, TRC

Product Code. 30450003
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450003 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450003 Supplier Toronto Research Chemicals Supplier No. TRCV11500010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Velneprit
CAS 342577-38-2
Molecular Formula C17H24F3N3O3S
Quantity 10 mg
Synonym N-[5-(Trifluoromethyl)pyridin-2-yl]-trans-4-(tert-butylsulfonylamino)cyclohexane-1-carboxamide, S 2367
InChI Formula InChI=1S/C17H24F3N3O3S/c1-16(2,3)27(25,26)23-13-7-4-11(5-8-13)15(24)22-14-9-6-12(10-21-14)17(18,19)20/h6,9-11,13,23H,4-5,7-8H2,1-3H3,(H,21,22,24)
SMILES CC(C)(S(=O)(N[C@H]1CC[C@H](C(NC2=NC=C(C(F)(F)F)C=C2)=O)CC1)=O)C
Molecular Weight (g/mol) 407.45
Formula Weight 407.149
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.