missing translation for 'onlineSavingsMsg'
Learn More

Velneperit, MedChemExpress

Product Code. p-200101728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496058 10 mg 10mg
16482197 50 mg 50mg
16458667 5 mg 5mg
16416466 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496058 Supplier Medchem Express Supplier No. HY14423

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Velneperit (S-2367) is a novel neuropeptide Y (NPY) Y5 receptor antagonist.

Velneperit (S-2367) is a novel neuropeptide Y (NPY) Y5 receptor antagonist.

Specifications

Chemical Name or Material Velneperit
CAS 342577-38-2
Color White
Purity Grade Notes Research
Molecular Formula C17H24F3N3O3S
Quantity 10 mg
Synonym S2367
Solubility Information DMSO : ≥ 30 mg/mL (73.63 mM)
SMILES FC(F)(F)C1=CN=C(NC([C@H]2CC[C@H](NS(=O)(C(C)(C)C)=O)CC2)=O)C=C1
Molecular Weight (g/mol) 407.45
Formula Weight 407.45
Percent Purity 99.5%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.