Learn More
Velneperit, MedChemExpress
Product Code. 16496058
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16496058
Brand: Medchem Express HY14423
MedChemExpress Velneperit (S-2367) is a novel neuropeptide Y (NPY) Y5 receptor antagonist.
Velneperit (S-2367) is a novel neuropeptide Y (NPY) Y5 receptor antagonist.
Specifications
Chemical Name or Material Velneperit
CAS 342577-38-2
Color White
Purity Grade Notes Research
Molecular Formula C17H24F3N3O3S
Quantity 10 mg
Synonym S2367
Solubility Information DMSO : ≥ 30 mg/mL (73.63 mM)
SMILES FC(F)(F)C1=CN=C(NC([C@H]2CC[C@H](NS(=O)(C(C)(C)C)=O)CC2)=O)C=C1
Molecular Weight (g/mol) 407.45
Formula Weight 407.45
Percent Purity 99.5%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism