missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR-1/FLT-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | VEGFR-1/FLT-1 |
---|---|
Concentration | 0.26 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882034
|
Proteintech
13687-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16872034
|
Proteintech
13687-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
VEGFR 1 is a cell membrane receptor kinase. It has a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis.Specifications
VEGFR-1/FLT-1 | |
Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P17948, P35969, P53767 | |
14254, 2321, 54251 | |
VEGFR-1/FLT-1 Fusion Protein Ag4558 | |
Primary | |
-20°C | |
FLT1 |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FLT, FLT 1, FLT1, FLT-1, Fms like tyrosine kinase 1, FRT, sflt1, Tyrosine protein kinase FRT, VEGF receptor 1, VEGFR 1, VEGFR1, VEGFR-1 | |
Flt1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |