missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFR-1/FLT-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13687-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
VEGFR 1 is a cell membrane receptor kinase. It has a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis.Specifications
VEGFR-1/FLT-1 | |
Polyclonal | |
Unconjugated | |
FLT1 | |
FLT, FLT 1, FLT1, FLT-1, Fms like tyrosine kinase 1, FRT, sflt1, Tyrosine protein kinase FRT, VEGF receptor 1, VEGFR 1, VEGFR1, VEGFR-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14254, 2321, 54251 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17948, P35969, P53767 | |
Flt1 | |
VEGFR-1/FLT-1 Fusion Protein Ag4558 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |