missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGF 121 Antibody (064), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89193MFV450
This item is not returnable.
View return policy
Description
VEGF 121 Monoclonal antibody specifically detects VEGF 121 in Human samples. It is validated for ELISA, ELISA
Specifications
| VEGF 121 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA | |
| 064 | |
| ELISA, Sandwich ELISA Detection | |
| MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGF 121 (Uniprot#: P15692-9; Met1-Arg147). | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins | |
| 7422 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction