missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-Cadherin Rat anti-Mouse, Alexa Fluor 405, Clone: 162709, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1002V-100UG
This item is not returnable.
View return policy
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Mouse samples. It is validated for Flow Cytometry.Specifications
| VE-Cadherin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| 2208309A | |
| CDH5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse VE-Cadherin Asp46-Gln592 (Gly67 del, Ile69Asp, Lys70Gln) Accession # 2208309A | |
| RUO | |
| 1003 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 162709 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse VE-Cadherin in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cadherin-8, rhCadherin-17, rhN-Cadherin, recombinant mouse (rm) E-Cadherin, or rmP-Cadherin is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction