Learn More
VACUUBRAND™ VARIO™ Chemistry Pumping Unit, PC 3002 VARIO Select
Product Code. 16201782
Change view
Click to view available options
Plug Type:
CEE
IEC Connector
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type unitSize
16201782 IEC Connector Each
15848797 CEE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16201782 Supplier VACUUBRAND™ Supplier No. 20733574
Only null left
Add to basket
Add to basket
Provides pinpoint vacuum control by adjusting the motor speed. The automatic evaporation mode is perfect for controlling applications such as rotary evaporation. A variety of condensers are available to suit a range of needs.
  • VACUU·SELECT Vacuum Controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Automatic boiling point detection and adjustment of the vacuum for short process times
  • Compact and efficient, good ultimate vacuum even with open gas ballast valve
  • Very quiet, low-vibration operation and extra long service intervals
  • Excellent environmental compatibility through low energy consumption and efficient solvent recovery
  • PC 3002 VARIO™ is suitable for work with most common solvents
  • Use manual set-point-control for simple processes, run fully automated distillations, or create unique applications with simple drag-and-drop editing
  • VACUU·SELECT Controller detects solvent boiling and automatically adjusts the pump's motor speed to maintain process control during solvent evaporation
  • Glass inlet separator with a robust protective coating, prevents particles and liquid droplets from entering the pump, preventing damage and extending pump life
  • Touch screen display can be operated even when wearing safety gloves
  • Choose from 14 languages
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 7 Mbar (Without Gas Ballast), 12 Mbar (With Gas Ballast)
Control VACUU·SELECT
Shipping Weight (Metric) 29 kg
Includes Ready-to-Use Pumping Unit (Completely Mounted with Manual)
Motor Speed 30 to 2400 min-1
Certifications/Compliance CE-certification; II 3/-G IIC T3 x Internal Atm. Only
Dimensions (L x W x H) 419 x 243 x 457 mm
Description Peltronic™ Vapor Condenser works without any cooling media
Material PTFE (Partly Carbon Reinforced), ETFE/ECTFE (Partly Carbon Fiber Reinforced), FFKM
Model PC 3002 VARIO Select EK Peltronic
Noise Level 43 dBA
Plug Type IEC Connector
Wattage 0.53 kW
Weight (Metric) 21.5 kg
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Flow Rate 2.8 m³/hr.
Frequency 50/60 Hz
Pump Type Diaphragm Pump
Enclosure Rating IP40
No. of Pump Heads 2
Applications Intelligent Vacuum with VARIO™-control, Rotary Evaporator/Parallel Evaporator, Vacuum Concentrators, Gel Dryer
No. of Stages 2
Maximum Pressure 1.1 bar
Voltage 200/230 V