missing translation for 'onlineSavingsMsg'
Learn More

Varenicline Tartrate, MedChemExpress

Product Code. p-200099318
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485587

Brand: Medchem Express HY10021

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Varenicline Tartrate(CP 526555;Champix) is a nicotinic receptor partial agonist; it stimulates nicotine receptors more weakly than nicotine itself does.

Varenicline Tartrate(CP 526555

Specifications

Chemical Name or Material Varenicline Tartrate
CAS 375815-87-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H19N3O6
Quantity 5 mg
Synonym CP 526555-18
Solubility Information H2O : 20 mg/mL (55.35 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 14.29 mg/mL (39.55 mM; Need ultrasonic)
SMILES O=C(O)[C@H](O)[C@@H](O)C(O)=O.C1(C2CC3CNC2)=C3C=C(N=CC=N4)C4=C1
Molecular Weight (g/mol) 361.35
Formula Weight 361.35
Percent Purity 97.67%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.