Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Bottles and Carboys
Click to view available options
Size:
51 mm
GL38
Nalgene 38-430
Wako 38 mm
Material:
PTFE
Polypropylene
Unit Size:
Each
Description
Various cap sizes available to fit your solvent container
Warranty
None
Specifications
Specifications
| For Use With (Equipment) | Solvent-Compatible Containers |
| Material | Polypropylene |
| Size | Wako 38 mm |
| Thread Size (English) | 1 x 1/2 in. NPT(F), 3 x 1/4 in. NPT(F), 6 x 1/4 in.-28 UN(F)(F) |
| For Use With (Application) | Solvent Safety |
| Type | Solvent Waste Kit |