Learn More
Testo™ Vane Anemometer with Humidity Measurement
Product Code. 10233183
Change view
Click to view available options
Dimensions (L x W x H):
133 x 46 x 25 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
10233183 133 x 46 x 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10233183 Supplier Testo™ Supplier No. 0560 4102
Specifications
Type Vane Anemometer
Accuracy ±0.5 m/sec.
Dimensions (L x W x H) 133 x 46 x 25 mm
For Use With (Application) Measures air flow, air temperature, and humidity
Temperature (Metric) -20 to +70°C