missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vancomycin hydrochloride, MedChemExpress
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Description
Vancomycin hydrochloride is an antibiotic for the treatment of bacterial infections. It acts by inhibiting the second stage of cell wall synthesis of susceptible bacteria. Vancomycin also alters the permeability of the cell membrane and selectively inhibits ribonucleic acid synthesis.
Specifications
Specifications
| Chemical Name or Material | Vancomycin hydrochloride |
| Physical Form | Solid |
| CAS | 1404-93-9 |
| Color | Light Nude |
| Quantity | 250 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C66H76Cl3N9O24 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | H2O : 33.33 mg/mL (22.43 mM; Need ultrasonic) ∣DMSO : 24 mg/mL (16.15 mM; Need ultrasonic and warming) |
| SMILES | ClC1=CC([C@H]([C@@H](NC([C@H](NC)CC(C)C)=O)C(N[C@@H](CC(N)=O)C(N[C@@]2([H])C(N[C@]3([H])C4=CC(C5=C(C=C(O)C=C5O)[C@](C(O)=O)([H])NC([C@](NC3=O)([H])[C@H](O)C6=CC=C7C(Cl)=C6)=O)=C(O)C=C4)=O)=O)=O)O)=CC=C1OC8=C(C(O7)=CC2=C8)O[C@H]9[C@@H]([C@H]([C@H](O)[C@@H](CO)O9)O)O[C@@]%10([H])C[C@](N)([C@H](O)[C@H](C)O%10)C.Cl |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction