Learn More
Vanadium Standard: V @ 1000 μg/g in Hydrocarbon Oil, VHG™
Product Code. 17932816
Change view
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932816 50 g 50g
17900386 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17932816 Supplier VHG Labs Supplier No. VHGOV100050G
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Vanadium Standard: V @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Vanadium
CAS 7440-62-2
Methods ICP/XRF/RDE
Analyte or Component Names Vanadium
Shipping Condition Room Temperature
Molecular Formula V
Synonym Vanadium dust, Vanadium, elemental, Vanadium (III)
InChI Formula InChI=1S/V
SMILES [V]
IUPAC Name vanadium
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 50.94
Quantity 50 g
Formula Weight 50.944 g/mol