Learn More
Gibco™ Value 10 Pack EMEM
Click to view available options
Quantity:
500 mL
Unit Size:
Pack of 10
Description
Contains phenol red, nonessential amino acids, 1 mM sodium pyruvate, 2 mM L-glutamine, 1 g/L glucose, and 1.5 g/L sodium bicarbonate.
Specifications
Specifications
| Content And Storage | Protect from light, 2°C to 8°C |
| Concentration | 10X |
| Form | Liquid |
| Product Type | Basal Media |
| For Use With (Application) | Cell Culture |
| Quantity | 500 mL |