missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Val-cit-PAB-OH

Product Code. 16421636
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 16421636

Brand: MedChemExpress HY12362

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Val-cit-PAB-OH is a cleavable ADC linker.

Specifications

Purity 98.8%
CAS 159857-79-1
Content And Storage 4°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 379.45 g/mol
Product Type Linker
Quantity 500 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 100 mg/mL (263.54 mM)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.