Learn More
Memmert™ Vacuum Connecting Hose 3M W/Connection Accessories (Not Required With Pump Module), For Vo29
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
Cooled vacuum drying oven Vocool
Unit Size:
Each
Description
- Vacuum connecting hose from oven to Memmert pump, 3m
- Includes optimized connection accessories
- Partially stainless steel
Specifications
Specifications
| For Use With (Equipment) | Cooled vacuum drying oven Vocool |
| Type | Vacuum Connecting Hose |
| Material | Stainless Steel |
| Length (Metric) | 300 cm |