Learn More
VACUUBRAND™ VACUU·LAN™ Manual Flow Control/Vacuum Gauge For VCL RMS
Description
The VCL RMS is an add-on module without mounting base. Based on a B1 module body (with DN 6/10 hose nozzle), it is used to add manual flow control and a vacuum gauge to mounting bases that are already in place. This module is convenient when mounted on side panels inside or outside an fume hood. The VCL RMS provides reliable and precise manual vacuum control;the manometer offers an analog vacuum display.The manual flow control and vacuum indication module is leak tested. (Image rotated 180° to show end that mounts on mounting base.)
Specifications
Specifications
| For Use With | For VCL RMS |
| Quantity | 1 Each |
| Type | Manual Flow Control/Vacuum Gauge |