missing translation for 'onlineSavingsMsg'
Learn More
Learn More
V12 Wear Metals Standard: 500 μg/g Al, Cr, Cu, Fe, Pb, Mg, Ni, Si, Ag, Na, Sn, Ti in Hydrocarbon Oil, VHG™
Description
V12 Wear Metals Standard: 500 μg/g Al, Cr, Cu, Fe, Pb, Mg, Ni, Si, Ag, Na, Sn, Ti in Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium |
| CAS | 7429-90-5, 7440-47-3, 7440-50-8, 7439-89-6, 7439-92-1, 7439-95-4 |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hydrocarbon Oil |
| Quantity | 50 g |
| Concentration or Composition (by Analyte or Components) | Aluminium 500 μg/g/Chromium 500 μg/g/Copper 500 μg/g/Iron 500 μg/g/Lead 500 μg/g/Magnesium 500 μg/g/Nickel 500 μg/g/Silicon 500 μg/g/Silver 500 μg/g/Sodium 500 μg/g/Tin 500 μg/g/Titanium 500 μg/g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?