missing translation for 'onlineSavingsMsg'
Learn More

V12 Wear Metals Standard: 300 μg/g Al, Cr, Cu, Fe, Pb, Mg, Ni, Si, Ag, Na, Sn, Ti in Hydrocarbon Oil, VHG™

Product Code. 30003196
Change view
Click to view available options
Quantity:
50 g
100 g
200 g
Unit Size:
100g
200g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003196 200 g 200g
30003023 50 g 50g
30002989 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003196 Supplier VHG Labs Supplier No. VHGV12300200G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

V12 Wear Metals Standard: 300 μg/g Al, Cr, Cu, Fe, Pb, Mg, Ni, Si, Ag, Na, Sn, Ti in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium
CAS 7429-90-5, 7440-47-3, 7440-50-8, 7439-89-6, 7439-92-1, 7439-95-4
Methods ICP/XRF/RDE
Analyte or Component Names Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Hydrocarbon Oil
Quantity 200 g
Concentration or Composition (by Analyte or Components) Aluminium 300 μg/g/Chromium 300 μg/g/Copper 300 μg/g/Iron 300 μg/g/Lead 300 μg/g/Magnesium 300 μg/g/Nickel 300 μg/g/Silicon 300 μg/g/Silver 300 μg/g/Sodium 300 μg/g/Tin 300 μg/g/Titanium 300 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.