missing translation for 'onlineSavingsMsg'
Learn More
Learn More
V12 Wear Metals Standard: 100 μg/g Al, Cr, Cu, Fe, Pb, Mg, Ni, Si, Ag, Na, Sn, Ti in Hydrocarbon Oil, VHG™
Click to view available options
Quantity:
50 g
100 g
200 g
Unit Size:
100g
200g
50g
Description
V12 Wear Metals Standard: 100 μg/g Al, Cr, Cu, Fe, Pb, Mg, Ni, Si, Ag, Na, Sn, Ti in Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium |
| CAS | 7429-90-5, 7440-47-3, 7440-50-8, 7439-89-6, 7439-92-1, 7439-95-4 |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hydrocarbon Oil |
| Quantity | 200 g |
| Concentration or Composition (by Analyte or Components) | Aluminium 100 μg/g/Chromium 100 μg/g/Copper 100 μg/g/Iron 100 μg/g/Lead 100 μg/g/Magnesium 100 μg/g/Nickel 100 μg/g/Silicon 100 μg/g/Silver 100 μg/g/Sodium 100 μg/g/Tin 100 μg/g/Titanium 100 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction