missing translation for 'onlineSavingsMsg'
Learn More

V12 Wear Metals Standard: 10 μg/g Al, Cr, Cu, Fe, Pb, Mg, Ni, Si, Ag, Na, Sn, Ti in Hydrocarbon Oil, VHG™

Product Code. 17933826
Change view
Click to view available options
Quantity:
50 g
100 g
200 g
Unit Size:
100g
200g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933826 100 g 100g
17910476 50 g 50g
17914296 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933826 Supplier VHG Labs Supplier No. VHGV1210100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

V12 Wear Metals Standard: 10 μg/g Al, Cr, Cu, Fe, Pb, Mg, Ni, Si, Ag, Na, Sn, Ti in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium
CAS 7429-90-5, 7440-47-3, 7440-50-8, 7439-89-6, 7439-92-1, 7439-95-4
Methods ICP/XRF/RDE
Analyte or Component Names Aluminium/Chromium/Copper/Iron/Lead/Magnesium/Nickel/Silicon/Silver/Sodium/Tin/Titanium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Hydrocarbon Oil
Quantity 100 g
Concentration or Composition (by Analyte or Components) Aluminium 10 μg/g/Chromium 10 μg/g/Copper 10 μg/g/Iron 10 μg/g/Lead 10 μg/g/Magnesium 10 μg/g/Nickel 10 μg/g/Silicon 10 μg/g/Silver 10 μg/g/Sodium 10 μg/g/Tin 10 μg/g/Titanium 10 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.