missing translation for 'onlineSavingsMsg'
Learn More

V-9302, MedChemExpress

Product Code. 16460967
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460967 50 mg 50mg
16454678 10 mg 10mg
16413196 25 mg 25mg
16407397 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16460967 Supplier Medchem Express Supplier No. HY112683

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress V-9302 is a competitive antagonist of transmembrane glutamine flux. V-9302 selectively and potently targets the amino acid transporter ASCT2 (SLC1A5) not ASCT1. V-9302 inhibits ASCT2-mediated glutamine uptake (IC50=9.6 μM) in HEK-293 cells.

V-9302 is a competitive antagonist of transmembrane glutamine flux. V-9302 selectively and potently targets the amino acid transporter ASCT2 (SLC1A5) not ASCT1. V-9302 inhibits ASCT2-mediated glutamine uptake (IC50=9.6 μM) in HEK-293 cells.

Specifications

Chemical Name or Material V-9302
CAS 1855871-76-9
Color White
Purity Grade Notes Research
Molecular Formula C34H38N2O4
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (46.41 mM; ultrasonic and warming and heat to 80°C) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(O)[C@@H](N)CCN(CC1=CC=CC=C1OCC2=CC=CC(C)=C2)CC3=CC=CC=C3OCC4=CC=CC(C)=C4
Molecular Weight (g/mol) 538.68
Formula Weight 538.68
Percent Purity 97.36%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.