missing translation for 'onlineSavingsMsg'
Learn More

Vβ 5.1, 5.2 T-Cell Receptor Mouse anti-Mouse, BUV661, Clone: MR9-4, BD Biosciences™

Product Code. 17221725
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17221725 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17221725 Supplier BD Biosciences Supplier No. 750367

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Vβ 5.1, 5.2 T-Cell Receptor Monoclonal antibody specifically detects Vβ 5.1, 5.2 T-Cell Receptor in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vβ 5.1, 5.2 T-Cell Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone MR9-4
Conjugate Brilliant Ultraviolet 661
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TCR Vβ5.1/5.2; Vβ5 T-cell receptor; T cell receptor beta 5
Host Species Mouse
Immunogen Mouse T-Cell Hybridoma 2HB51.8
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.