missing translation for 'onlineSavingsMsg'
Learn More

Uzansertib phosphate, MedChemExpress

Product Code. 16462796
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16462796

Brand: Medchem Express HY101870B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Uzansertib (INCB053914) phosphate is an orally active, ATP-competitive pan-PIM kinase inhibitor with IC50s of 0.24 nM, 30 nM, 0.12 nM for PIM1, PIM2, PIM3, respectively. Uzansertib phosphate has broad anti-proliferative activity against a variety of hematologic tumor cell lines.

Uzansertib (INCB053914) phosphate is an orally active, ATP-competitive pan-PIM kinase inhibitor with IC50s of 0.24 nM, 30 nM, 0.12 nM for PIM1, PIM2, PIM3, respectively. Uzansertib phosphate has broad anti-proliferative activity against a variety of hematologic tumor cell lines.

Specifications

Chemical Name or Material Uzansertib phosphate
CAS 2088852-47-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H29F3N5O7P
Quantity 25 mg
Synonym INCB053914 phosphate
Solubility Information DMSO : 5 mg/mL (8.18 mM; Need ultrasonic and warming)
SMILES O=C(C1=NC(C2=C(F)C=CC=C2F)=C(F)C=C1)NC3=CN=C4C(CC[C@H]4O)=C3N5C[C@@H](N)[C@H](O)[C@@H](C)C5.OP(O)(O)=O
Molecular Weight (g/mol) 611.51
Formula Weight 611.51
Percent Purity 98.33%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.