Learn More
UVP Replacement UV Light Tubes
Product Code. 12842989
Change view
Click to view available options
Wavelength:
302 nm
365 nm
Wattage:
6 W
15 W
20 W
40 W
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Wattage unitSize
12842989 302 nm 6 W Each
12813858 365 nm 40 W Each
12893848 365 nm 20 W Each
12883918 365 nm 15 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12842989 Supplier UVP Supplier No. 34004401
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Replacement UV light tubes for 4, 6, 15, 20 and 40 watt ultraviolet lamps and transilluminators
Specifications
Wattage 6 W
Wavelength 302 nm
For Use With (Equipment) EL-series UV lamp, 3UV™ lamp, handheld UV lamp, mid size UV viewing cabinets, benchtop UV transilluminators
Length (English) 8.3 in.
Length (Metric) 210.8 mm
Packaging Quantity 1 Ea.