Learn More
UVP Lamp Stands
Product Code. 11786812
Change view
Click to view available options
Model:
J-124
J-129
J-138
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
11786812 J-129 Each
12611732 J-138 Each
11718231 J-124 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11786812 Supplier UVP Supplier No. 98001603
Allows easy and comfortable hands free operation of UV lamps
Allows easy and comfortable hands free operation of UV lamps
  • Housing: Varies from metal housing with scratch resistant powder coat to plastic housing
  • Allows hands free operation of the lamp
Specifications
Dimensions (L x W x H) 13 x 3 x 10.74 in. (330 x 76 x 273 mm)
For Use With (Equipment) EL-series UV lamp
Height (English) 10.74 in.
Height (Metric) 273 mm
Length (English) 13 in.
Length (Metric) 330 mm
Model J-129
Packaging Quantity 1 Ea.
Width (English) 3 in.
Width (Metric) 76 mm
Product Type Lamp Stand