missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Standard, Bulk
GREENER_CHOICE

Product Code. p-7146922
Change view
Click to view available options
Volume (Metric):
250 μL
1000 μL
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
30216564 250 μL Pack of 1000
30216538 1000 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30216564 Supplier BIOTIX™ Supplier No. 63300079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Top quality pipette tips in resealable bags. Validated for fit and accuracy on universal pipettes.

  • Validated for fit and accuracy on standard pipettes like Eppendorf™, Gilson™ and Biohit™
  • Low binding for Protein, DNA and nanoparticles
  • Re-sealable bag
  • Tips must be racked by hand
  • Certifications

    ISO 9001

    TRUSTED_SUSTAINABILITY

    Specifications

    Description 250 μL, Bulk
    Volume (Metric) 250 μL
    Filtered Non-filtered
    Sterility Non-sterile
    Color Clear
    Autoclavable Not Autoclavable
    Model Universal
    Length (Metric) 59.54 mm
    Certifications/Compliance ISO 9001
    Clean Claims Fully Recyclable
    For Use With (Application) Low binding for Protein, DNA, and nanoparticles
    Format Bulk
    Tip Style Ergonomic
    Quantity 1000 tips/bag
    Product Type Pipette Tips
    Disposable Single-use
    For Use With (Equipment) Universal Fit
    Graduated Yes
    Product Line uTip
    Material Polypropylene
    Shape Conical
    Graduations 10 μL, 50 μL, 100 μL, 200 μL
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.