missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP7-IN-1, MedChemExpress
Description
USP7-IN-1 is a selective and reversible inhibitor of ubiquitin-specific protease 7 (USP7), with an IC50 of 77 μM, and can be used for the research of cancer.
Specifications
Specifications
| Chemical Name or Material | USP7-IN-1 |
| Physical Form | Solid |
| CAS | 1381291-36-6 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C23H24ClN3O3 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : ≥ 40 mg/mL (93.92 mM) |
| SMILES | O=C1N(CC2(O)CCN(C(CCC3=CC=CC=C3)=O)CC2)C=NC4=C1C=CC(Cl)=C4 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?