missing translation for 'onlineSavingsMsg'
Learn More

USP44 Monoclonal Antibody (OTI1D6), TrueMAB™, OriGene

Product Code. 17590343
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17590343 30 μL 30µL
17570343 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17590343 Supplier Origene Technologies Supplier No. TA801910S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

USP44 Monoclonal antibody specifically detects USP44 in Human samples. It is validated for Western Blot

Specifications

Antigen USP44
Applications Western Blot
Classification Monoclonal
Clone OTI1D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene USP44
Gene Accession No. Q9H0E7
Gene Alias Deubiquitinating enzyme 44; E430004F17Rik; hypothetical protein MGC55661; inactive deubiquitinating enzyme 44; inactive ubiquitin carboxyl-terminal hydrolase 44; inactive ubiquitin thioesterase 44; inactive ubiquitin-specific-processing protease 44; LOW QUALITY PROTEIN: ubiquitin carboxyl-terminal hydrolase 44; ubiquitin carboxyl-terminal hydrolase 44; ubiquitin specific peptidase 44; ubiquitin specific peptidase 49 pseudogene; ubiquitin specific protease 44; ubiquitin thioesterase 44; ubiquitin thiolesterase 44; ubiquitin-specific-processing protease 44; USP44; zgc:55661
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 420-712 of human USP44 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84101
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.