Learn More
USP28 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17707-1-AP-150UL
This item is not returnable.
View return policy
Description
WNK is a serine/threonine kinase that plays an important role in the regulation of electrolyte homeostasis, cell signaling, survival, and proliferation. Also, acts as an activator and inhibitor of sodium-coupled chloride cotransporters and potassium-coupled chloride cotransporters respectively. Activates SCNN1A, SCNN1B, SCNN1D, and SGK1. WNK localizes to a segment of kidney involved in salt, potassium, and pH homeostasis, the distal nephron. Mutations in the WNK gene result in Pseudohypaldosteronism and Neuropathy.Specifications
| USP28 | |
| Polyclonal | |
| Unconjugated | |
| USP28 | |
| Deubiquitinating enzyme 28, KIAA1515, Ubiquitin thiolesterase 28, USP28 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 235323, 315639, 57646 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D3ZJ96, Q5I043, Q96RU2 | |
| Usp28 | |
| USP28 Fusion Protein Ag12003 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |