missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP/ICH Oral Target Elements Standard C R2: Au @ 300; Ir, Os, Pd, Pt, Rh, Ru @ 100 μg/mL in 15% HCl
Specifications
Specifications
| Chemical Name or Material | Gold∣Iridium∣Osmium∣Palladium∣Platinum∣Rhodium∣Ruthenium |
| CAS | 7440-57-5, 7439-88-5, 4-2-7440, 5-3-7440, 6-4-7440, 7440-16-6 |
| Analyte or Component Names | Gold∣Iridium∣Osmium∣Palladium∣Platinum∣Rhodium∣Ruthenium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Solution Type | Mixture |
| Element | Multi-element |
| Solvent or Matrix | Dilute HCl |
| Industry Type | Industrial QC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?