Learn More
Brady™ USB Cable
Product Code. 11579313
Click to view available options
Unit Size:
Each
Product Code. 11579313
Brand: Brady™ 076805
Only null left
Add to basket
Add to basket
USB Cable for use with Global Mark Printers.
  • For Use with: Global Mark Printer
Specifications
Product Type USB Cable
For Use With (Equipment) Wraptor A6500
Description USB Cable
Format Cable
Quantity 1 Ea.