missing translation for 'onlineSavingsMsg'
Learn More

Ursolic Acid, 98%, MP Biomedicals™

Product Code. 11479721 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11479721

Brand: MP Biomedicals 0215825390

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 77-52-1
Molecular Formula C30H48O3
Molecular Weight (g/mol) 456.71
MDL Number MFCD00009621
InChI Key WCGUUGGRBIKTOS-GPOJBZKASA-N
Synonym 3β-Hydroxy-12-ursen-28-ic acid
PubChem CID 129316832
IUPAC Name (1S,2R,4aS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-hydroxy-1,2,6a,6b,9,9,12a-heptamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid
SMILES C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(O)=O

Specifications

Assay Percent Range 0.98
Quantity 500 mg
Formula Weight 456.71
Physical Form Powder
Chemical Name or Material Ursolic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.