missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uridine-5'-diphosphogalactose Disodium Salt, ≥98%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| CAS | 137868-52-1 |
| Assay Percent Range | ≥98% |
| Molecular Formula | C15H22N2Na2O17P2 |
| MDL Number | MFCD00077895 |
| Quantity | 25 mg |
| Synonym | UDPG, UDP-Galactose |
| InChI Key | PKJQEQVCYGYYMM-UHFFFAOYNA-L |
| SMILES | [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O |
| IUPAC Name | disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate |
| Molecular Weight (g/mol) | 610.27 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction