missing translation for 'onlineSavingsMsg'
Learn More

Uridine-5'-Diphospho-galactose disodium salt, ≥98%, MP Biomedicals™

Product Code. 11432611 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 11432611

Brand: MP Biomedicals 0219590505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

UDP-Gal and its analogues are used to study the distribution, specificity and kinetics of galactosyltransferase(s)

TRUSTED_SUSTAINABILITY

Specifications

CAS 137868-52-1
Color White
Assay Percent Range ≥98%
Molecular Formula C15H22N2Na2O17P2
MDL Number MFCD00077895
Quantity 5 mg
Synonym udp-a-d-galactose disodium salt
InChI Key PKJQEQVCYGYYMM-UHFFFAOYNA-L
SMILES [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O
IUPAC Name disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
Molecular Weight (g/mol) 610.27
PubChem CID 126969254
Formula Weight 610.3
Physical Form Powder
Chemical Name or Material Uridine-5'-Diphospho-galactose disodium salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.