missing translation for 'onlineSavingsMsg'
Learn More

Urea-15N2, TRC

Product Code. 30133625
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133625 100 mg 100mg
30133700 250 mg 250mg
17959534 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133625 Supplier Toronto Research Chemicals Supplier No. TRCU822504100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2067-80-3
Molecular Formula CH415N2O
Molecular Weight (g/mol) 62.04
Synonym Aquacare-15N2, Aquadrate-15N2, B-I-K-15N2, Basodexan-15N2, Benural 70-15N2, Carbamide-15N2, Carbamimidic Acid-15N2, Carbonyl Diamide-15N2, Carmol 40-15N2, Keratinamin-15N2, Urepearl-15N2;
SMILES [15NH2]C([15NH2])=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i2+1,3+1
Formula Weight 62.0264
Chemical Name or Material Urea-15N2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.