missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UQCRQ Polyclonal Antibody
GREENER_CHOICE

Product Code. 15567012
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15567012 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15567012 Supplier Invitrogen™ Supplier No. PA545979

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: KGIPNVLRRI RESFFRVVPQ FVVFYLIYTW GTEEFERSKR KNPAAYENDK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 92%; Rat: 92%.

This gene encodes a ubiquinone-binding protein of low molecular mass. This protein is a small core-associated protein and a subunit of ubiquinol-cytochrome c reductase complex III, which is part of the mitochondrial respiratory chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRQ
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UQCRQ
Gene Accession No. O14949
Gene Alias 1100001F06Rik; 1500040F11Rik; 5830407L17Rik; 9.5kDa; AA959903; c1502; Complex III subunit 8; complex III subunit VII; complex III subunit VIII; Cytochrome b-c1 complex subunit 8; low molecular mass ubiquinone-binding protein; low molecular mass ubiquinone-binding protein (9.5kD); low molecular mass ubiquinone-binding protein (Qpc-pending); MC3DN4; QCR8; QPC; QP-C; transcript under zygotic control; ubiquinol-cytochrome c reductase binding protein; ubiquinol-cytochrome c reductase complex 9.5 kDa protein; ubiquinol-cytochrome c reductase complex III subunit VII; ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; ubiquinol-cytochrome c reductase, complex III subunit VII; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa; unknown; UQCR7; Uqcrb; UQCRQ
Gene Symbols UQCRQ
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human UQCRQ.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27089
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.