missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCRC2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97364AF647
This item is not returnable.
View return policy
Description
UQCRC2 Polyclonal specifically detects UQCRC2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| UQCRC2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 7385 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| Complex III subunit 2, Core protein II, cytochrome b-c1 complex subunit 2, mitochondrial, QCR2, ubiquinol-cytochrome c reductase core protein II, Ubiquinol-cytochrome-c reductase complex core protein 2, UQCR2 | |
| Produced in rabbits immunized with E. coli-derived Human UQCRC2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction