Learn More
Fisherbrand™ Upright ULT Freezer Sliding Tray Racks
Product Code. 15709569 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
12 Boxes (2 in.)
15 (2in.) boxes
15 (3 in.) boxes
16 (2in.) boxes
20 (2in.) boxes
25 (2in.) boxes
28 (2in.) boxes
Dimensions (L x W x H):
419 x 140 x 225 mm
419 x 140 x 280 mm
559 x 140 x 225 mm
559 x 140 x 280 mm
559 x 140 x 391 mm
675 x 140 x 280 mm
679 x 140 x 225 mm
679 x 140 x 241 mm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Dimensions (L x W x H) unitSize
15709569 15 (3 in.) boxes 679 x 140 x 241 mm Each
15719569 20 (2in.) boxes 679 x 140 x 225 mm Each
15729569 25 (2in.) boxes 675 x 140 x 280 mm Each
15759559 12 Boxes (2 in.) 419 x 140 x 225 mm Each
15769559 15 (2in.) boxes 419 x 140 x 280 mm Each
15779559 16 (2in.) boxes 559 x 140 x 225 mm Each
15789559 20 (2in.) boxes 559 x 140 x 280 mm Each
15799559 28 (2in.) boxes 559 x 140 x 391 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15709569
Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of upright ULT freezers.
  • Holds standard cryoboxes
  • Made of high quality stainless steel
  • Sliding trays provide more flexibility and easy access to the boxes
Specifications
Holds 15 (3 in.) boxes
Material Stainless Steel
For Use With (Application) Standard 3 in. Cryoboxes
For Use With (Equipment) Upright ULT Freezer
Dimensions (L x W x H) 679 x 140 x 241 mm
Product Type Sliding Tray Rack