missing translation for 'onlineSavingsMsg'
Learn More

Upamostat, MedChemExpress

Product Code. p-200097017
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483526

Brand: Medchem Express HY16511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Upamostat (WX-671) is a serine protease inhibitor. Upamostat is the orally available prodrug of the WX-UK1, which is a urokinase plasminogen activator (uPA) inhibitor.

Upamostat (WX-671) is a serine protease inhibitor. Upamostat is the orally available prodrug of the WX-UK1, which is a urokinase plasminogen activator (uPA) inhibitor.

Specifications

Chemical Name or Material Upamostat
CAS 590368-25-5
Color White
Purity Grade Notes Research
Molecular Formula C32H47N5O6S
Quantity 25 mg
Synonym WX-671
Solubility Information DMSO : ≥ 250 mg/mL (396.95 mM)
SMILES O=C(N1CCN(C([C@@H](NS(=O)(C2=C(C(C)C)C=C(C(C)C)C=C2C(C)C)=O)CC3=CC=CC(C(NO)=N)=C3)=O)CC1)OCC
Molecular Weight (g/mol) 629.81
Formula Weight 629.81
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.