Learn More
Thermo Scientific™ Accessories for Digital Mini Rotators
Shop All Thermo Scientific Products
Click to view available options
Description:
1 L Flask Clamp
100/125 mL Flask Clamp
2 L Flask Clamp
250 mL Flask Clamp
50 mL Flask Clamp
500 mL Flask Clamp
Flat Platform w/ Rubber Mat
Sticky Mat, 14 x 14 cm
Tube support 1.5 mL
Tube support 15 mL
Tube support 50 mL
Universal (Aluminum) Platform
Universal Attachment
Dimensions (L x W):
59 x 38 mm
66 x 43 mm
85 x 55 mm
105 x 59 mm
130 x 77 mm
168 x 91 mm
197 x 86 mm
140 x 140 mm
262 x 112 mm
307 x 303 mm
310 x 284 mm
330 x 270 mm
missing translation for 'unitSize'
Each
Description
Assorted accessories for use with Digital Mini Rotators
Service Plan
Standard Thermo warranty guideline, provide repair service and free spare part within warranty
Services
2 years
Specifications
Specifications
| Product Type | Universal Platform |
| Certifications/Compliance | Certificate of Analysis (COA), Certificate of Conformance (COC) |
| Material | Aluminum |
| Dimensions (L x W) | 310 x 284 mm |
| Length (English) Platform | 12.21 in. |
| Length (Metric) Platform | 310 mm |
| Width (English) Platform | 11.18 in. |
| Width (Metric) Platform | 284 mm |
| Description | Universal (Aluminum) Platform |
| For Use With (Equipment) | Digital Mini Rotators, Rocker, Rotator, Mixer |
| Show More |